Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-PTPRA (Tyr798) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538323
Description
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP contains an extracellular domain, a single transmembrane segment and two tandem intracytoplasmic catalytic domains, and thus represents a receptor-type PTP. This PTP has been shown to dephosphorylate and activate Src family tyrosine kinases, and is implicated in the regulation of integrin signaling, cell adhesion and proliferation. Three alternatively spliced variants of this gene, which encode two distinct isoforms, have been reported.
Specifications
| Phospho-PTPRA (Tyr798) | |
| Polyclonal | |
| Unconjugated | |
| PTPRA | |
| PTPRA, HLPR, Hptp alpha, HEPTP, HPTPA, PTPA, Ptpalpha, PTPRL2, PTP alpha, PTPLCA-related phosphatase, R-PTP-alpha, RPTPA, Rptpalpha, Tyrosine phosphatase alpha, RPTP alpha, HPTPalpha, LRP, PTPase-alpha | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 19262, 25167, 5786 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P18433, P18052, Q03348 | |
| PTPRA | |
| A synthetic phosphopeptide derived from human PTPRA around the phosphorylation site of Tyr798 (S-D-YP-A-N) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction