Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-PTPRA (Tyr798) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986390
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6390 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6390 Supplier Bioss Supplier No. BS5179R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phosphorylation of receptors by protein kinases is a process that can be reversed by a group of enzymes called protein phosphatases. Coordinated control of kinases and phosphatases provides the cell with the capacity to rapidly switch between phosphorylated and dephosphorylated protein states in dynamic response to environmental stimuli. Activation of critical enzymes by kinase phosphorylation alone is not enough to provide adequate regulation - it is the combination with phosphatase dephosphorylation that effectively creates on/off switches to control cellular events. Errors in control, either through kinases or their counterpart phosphatases, can lead to unchecked cell growth attributable to human cancers and developmental disorders. Potential mechanisms to control dephosphorylation include changes in the expression of protein phosphatases, their subcellular localization, phosphorylation of phosphatase catalytic and regulatory subunits and regulation by endogenous phosphatase inhibitors. Most protein phosphatases are not stringently specific for their substrates. Consequently, changes in phosphatase activity may have a broad impact on dephosphorylation and turnover of phosphoproteins that are substrates for different kinases. This may be an important point of control to connect cellular circuitry of interrelated signaling pathways, and to synchronize physiological responses.

Specifications

Antigen Phospho-PTPRA (Tyr798)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Ptpra
Gene Accession No. P18433
Gene Alias HEPTP; HLPR; Hptp alpha; HPTPA; HPTPalpha; LCA-related phosphatase; Leukocyte common antigen-related peptide (protein tyrosine phosphate); Lrp; protein tyrosine phosphatase alpha; protein tyrosine phosphatase, receptor type A; protein tyrosine phosphatase, receptor type, A; protein tyrosine phosphatase, receptor type, alpha polypeptide; Protein-tyrosine phosphatase alpha; PTP alpha; PTPA; Ptpalpha; PTPase-alpha; PTPLCA-related phosphatase; PTPRA; PTPRL2; PTPTY-28; Receptor-type tyrosine-protein phosphatase alpha; RPTP alpha; RPTPA; Rptpalpha; R-PTP-alpha; Rptra; Rptralpha; tyrosine phosphatase alpha
Gene Symbols Ptpra
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human PTPRA around the phosphorylation site of Tyr798.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5786
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.