Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-PYK2 (Tyr579) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537573
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537573 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537573 Supplier Invitrogen™ Supplier No. PA537573
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control for Western blot is NIH-3T3 or HepG2 cells; suggested positive control for IHC is human brain tissue.

PTK2B (FAK2) is a cytoplasmic tyrosine kinase involved in calcium-induced regulation of ion channels and activation of the map kinase signaling pathway. PTK2B regulates reorganization of the actin cytoskeleton, cell polarization, cell migration, adhesion, spreading, and bone remodeling. FAK2 plays a role in the regulation of the humoral immune response and is required for normal levels of marginal B-cells in the spleen and normal migration of splenic B-cells. It functions in signaling downstream of integrin and collagen receptors, immune receptors, G-protein coupled receptors, cytokine, chemokine, and growth factor receptors, and mediates responses to cellular stress. Aberrant FAK2 expression may play a role in cancer cell proliferation, migration, and invasion in tumor formation and metastasis. Elevated expression has been seen in gliomas, hepatocellular carcinoma, lung cancer, and breast cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-PYK2 (Tyr579)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PTK2B
Gene Accession No. Q14289, Q9QVP9
Gene Alias CADTK; CAK beta; CAKB; CAKbeta; CAK-beta; calcium-dependent tyrosine kinase; Calcium-regulated non-receptor proline-rich tyrosine kinase; cell adhesion kinase beta; cellular adhesion kinase beta; E430023O05Rik; FADK; FADK 2; FADK2; FAK; FAK1; FAK2; focal adhesion kinase 2; FRNK; PKB; pp125FAK; proline-rich tyrosine kinase 2; protein kinase B; protein tyrosine kinase 2 beta; protein-tyrosine kinase 2-beta; PTK; PTK2 protein tyrosine kinase 2 beta; PTK2B; PTK2B protein tyrosine kinase 2 beta; PYK2; RAFTK; related adhesion focal tyrosine kinase
Gene Symbols PTK2B
Host Species Rabbit
Immunogen Peptide sequence around phosphorylation site of tyrosine 579(E-D-Y(p)-Y-K) derived from Human PYK2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19229, 2185
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.