Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phospho Rab1 (Thr75) Rabbit anti-Human, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABS213125UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS2131-25UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS2131-25UL Supplier MilliporeSigma Supplier No. ABS213125UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-phospho Rab1 (Thr75), Cat. No. ABS2131, is a rabbit polyclonal antibody that detects Ras-related protein Rab-1A and has been tested for use in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho Rab1 (Thr75)
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in PBS with 0.02% sodium azide.
Gene Alias YPT-1 related protein;Ras-related protein Rab-1A
Gene Symbols RAB1A;RAB1
Host Species Rabbit
Immunogen A linear peptide corresponding to 14 amino acids surrounding phosphorylated Thr75 from the N-terminal half of human Ras-related protein Rab-1A.
Molecular Weight of Antigen ∽48 kDa observed; 22.68 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects Ras-related protein Rab-1A (Rab1) in human cells. It targets an epitope within 14 amino acids surrounding phosphorylated Thr75.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.