Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-RAB29 (Ser72) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114630
Description
Antibody detects endogenous levels of RAB7L1 only when phosphorylated at Rab29.
Rab GTPase key regulator in vesicle trafficking. Essential for maintaining the integrity of the endosome-trans-Golgi network structure. Together with LRRK2, plays a role in the retrograde trafficking pathway for recycling proteins, such as mannose 6 phosphate receptor (M6PR), between lysosomes and the Golgi apparatus in a retromer-dependent manner. Regulates neuronal process morphology in the intact central nervous system (CNS). May play a role in the formation of typhoid toxin transport intermediates during Salmonella enterica serovar Typhi (S.Typhi) epithelial cell infection. {ECO:0000269; PubMed:22042847, ECO:0000269; PubMed:24788816}.
Specifications
Phospho-RAB29 (Ser72) | |
Polyclonal | |
Unconjugated | |
RAB29 | |
RAB29; RAB29, member RAS oncogene family; RAB7, member RAS oncogene family-like 1; RAB7L; RAB7L1; Rab-7-like protein 1; Ras-related GTP-binding protein Rab29; ras-related protein Rab-29; Ras-related protein Rab-7L1 | |
Rabbit | |
Sequential Chromatography | |
RUO | |
171122, 226422, 8934 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
O14966, Q63481, Q91YQ1 | |
RAB29 | |
A synthesized peptide derived from human Rab7L1 around the phosphorylation site of Rab29. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction