Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-RAD51 (Thr309) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986716
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6716 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6716 Supplier Bioss Supplier No. BS5677R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RAD51 plays a critical role in homologous strand exchange, a key step in DNA repair through homologous recobination. It binds to single and double-stranded DNA and exhibits DNA-dependent ATPase activity. RAD51 catalyzes the recognition of homology and strand exchange between homologous DNA partners to form a joint molecule between a processed DNA break and the repair template. It binds to a single-stranded DNA in an ATP-dependent manner to form nucleoprotein filaments which are essential for the homology search and strand exchange. Mutations in the gene can results in breast cancer, mirror movements 2 and fanconi anemia.

Specifications

Antigen Phospho-RAD51 (Thr309)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Rad51
Gene Accession No. Q06609
Gene Alias AV304093; BOS_10731; BRCA1/BRCA2-containing complex, subunit 5; BRCC5; DNA repair protein RAD51; DNA repair protein RAD51 homolog 1; FANCR; homolog to S.cerevisiae; HRAD51; HsRAD51; HsT16930; MRMV2; MUT5; Rad51; RAD51 homolog; RAD51 homolog (RecA homolog, E. coli); RAD51 homolog (S. cerevisiae); RAD51 homolog A; RAD51 recombinase; Rad51a; RECA; RecA, E. coli, homolog of; RecA-like protein; recombinase RAD51; recombination protein A; RGD1563603; wu:fb38e12; YER095W; zgc:77754
Gene Symbols Rad51
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human RAD51 around the phosphorylation site of Thr309 [GE (p-T)RI].
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5888
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.