Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-RAD9 (Ser328) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA512670
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA512670 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA512670 Supplier Invitrogen™ Supplier No. PA512670
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-RAD9 (Ser328) Polyclonal Antibody for DB, IHC (P), Western Blot

RAD9 is a component of the 9-1-1 cell cycle checkpoint response complex that plays a major role in DNA repair. The 9-1-1 complex is recruited to DNA lesions upon damage by the RAD17-replication factor C clamp loader complex. RAD9 acts as a sliding clamp platform on DNA for several proteins involved in long-patch base excision repair.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-RAD9 (Ser328)
Applications Dot Blot, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.32 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene RAD9A
Gene Accession No. Q99638
Gene Alias Cell cycle checkpoint control protein RAD9A; chromatin-binding protein RAD9; DNA repair exonuclease rad9 homolog A; DNA repair protein RAD9; hRAD9; mRAD9; QtsA-19913; Rad9; RAD9 checkpoint clamp component A; RAD9 homolog A; RAD9 homolog A (S. pombe); Rad9a; Rad9-like protein; rCG47324-like; unnamed protein product; YD9934.02C; YDR217C
Gene Symbols RAD9A
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S328 of human RAD9.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5883
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.