Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Raf-1 (S301) Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Supplier: R&D Systems PPS060
This item is not returnable.
View return policy
Description
Raf-1 Polyclonal specifically detects Raf-1 in Bovine, Canine, Chicken, Primate samples. It is validated for Western Blot.Specifications
Raf-1 (p ser301) | |
Polyclonal | |
Western Blot 1:1000 dilution | |
RAF1 | |
Phosphopeptide corresponding to amino acid residues surrounding the phospho-S301 of Raf-1 | |
100 μL | |
Primary | |
Human, mouse, rat, and Xenopus ∼73 kDa Raf-1 in Western blots. | |
For long-term storage, < -20degrees C is recommended. Product is stable at < -20degrees C for at least 1 year. |
Western Blot | |
Unconjugated | |
cRaf, c-Raf, c-Raf-1, CRAFraf proto-oncogene serine/threonine protein kinase, EC 2.7.11, EC 2.7.11.1, NS5, Proto-oncogene c-RAF, RAF, RAF proto-oncogene serine/threonine-protein kinase, Raf1, Raf-1Oncogene RAF1, v-Raf-1, v-raf-1 murine leukemia viral oncogene homolog 1 | |
Rabbit | |
Affinity Purified | |
RUO | |
5894 | |
Bovine, Canine, Chicken, Primate | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction