Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-RANBP3 (Ser126, Ser58) Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5104641
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104641 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104641 Supplier Thermo Scientific Supplier No. PA5104641

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of RanBP3 only when phosphorylated at Ser126/58.

Ran binding protein 3 (RanBP3) is a member of Ran GTPase binding proteins that is located at the nuclear pore complex (NPC) and plays an essential role in the transport between cytoplasm and nucleus. RANPB3 is expessed as two spliced forms, RanBP3a and RanBP3b, both of which contains two FXFG motifs, characteristic of a subgroup of nucleoporins in addition to nuclear localization signal in the amino-terminal region (amino acid residues 40-57) and a unique acidic carboxyl terminus which is integral to its switch function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-RANBP3 (Ser126, Ser58)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RANBP3
Gene Accession No. Q9CT10, Q9H6Z4
Gene Alias 2610024N24Rik; AA408221; RAN binding protein 3; RANB3; ran-binding protein 3; RAN-binding protein-3; Ranbp3
Gene Symbols RANBP3
Host Species Rabbit
Immunogen A synthesized peptide derived from human RANBP3(Accession Q9H6Z4), corresponding to amino acid residues around phosphorylated Ser126.
Purification Method sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 71810, 8498
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.