Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-RASGRF1 (Ser916) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537790
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537790 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537790 Supplier Invitrogen™ Supplier No. PA537790
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control for IHC is human brain tissue.

RASGRF1 is a guanine nucleotide exchange factor (GEF) similar to the Saccharomyces cerevisiae CDC25 gene product. Functional analysis has demonstrated that this protein stimulates the dissociation of GDP from RAS protein. Studies of the similar gene in mouse suggested that the Ras-GEF activity of this protein in brain can be activated by Ca2+ influx, muscarinic receptors, and G protein beta-gamma subunit. Mouse studies also indicated that the Ras-GEF signaling pathway mediated by this protein may be important for long-term memory.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-RASGRF1 (Ser916)
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RASGRF1
Gene Accession No. P27671, P28818, Q13972
Gene Alias AI844718; ASPP1; Cdc25; CDC25L; CDC25Mm; Gnrp; GRF1; GRF55; Grfbeta; guanine nucleotide exchange factor; guanine nucleotide-releasing factor 1; guanine nucleotide-releasing factor, 55 kD; guanine nucleotide-releasing protein; H-GRF55; KIAA0771; P140 Ras-GRF; p190; P190-A; p190RhoGEF; PP13187; PPP1R13B; Ras guanine release factor beta; Ras protein specific guanine nucleotide releasing factor 1; RAS protein-specific guanine nucleotide-releasing factor 1; Rasgrf1; ras-GRF1; Ras-specific guanine nucleotide-releasing factor 1; Ras-specific guanine nucleotide-releasing factor, CDC25 homolog; ras-specific nucleotide exchange factor CDC25
Gene Symbols RASGRF1
Host Species Rabbit
Immunogen Peptide sequence around phosphorylation site of Serine 916(N-E-G(p)-T P) derived from Human Ras-GRF1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192213, 19417, 5923
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.