Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-RCAN1 (Ser108) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564674
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564674 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564674 Supplier Invitrogen™ Supplier No. PA564674
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-RCAN1 (Ser108) Polyclonal Antibody detects endogenous levels of RCAN1 only when phosphorylated at Ser108.

The RCAN1 protein interacts with calcineurin A and inhibits calcineurin-dependent signaling pathways, possibly affecting central nervous system development. This gene is located in the minimal candidate region for the Down syndrome phenotype, and is overexpressed in the brain of Down syndrome fetuses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-RCAN1 (Ser108)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 30% glycerol, 0.87% NaCl and 0.01% sodium azide; pH 7.4
Gene Rcan1
Gene Accession No. P53805, Q6IN33, Q9JHG6
Gene Alias Adapt78; calcineurin inhibitor; calcipressin-1; calcium and oxidant-inducible mRNA; CALP1L; CSP1; Down syndrome candidate region 1; Down syndrome critical region gene 1; Down syndrome critical region homolog 1; Down syndrome critical region protein 1; Down syndrome critical region protein 1 homolog; DSC1; Dscr1; MCIP1; modulatory calcineurin-interacting protein 1; myocyte-enriched calcineurin-interacting protein 1; near DSCR proline-rich protein; Rcan1; RCN1; Regulator of calcineurin 1
Gene Symbols Rcan1
Host Species Rabbit
Immunogen A synthesized peptide derived from human DSCR1 (Phospho-Ser108).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1827, 266766, 54720
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.