Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-REC8 (Ser251) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987961
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7961 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7961 Supplier Bioss Supplier No. BS7726R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the kleisin family of SMC (structural maintenance of chromosome) protein partners. The protein localizes to the axial elements of chromosomes during meiosis in both oocytes and spermatocytes. In the mouse, the homologous protein is a key component of the meiotic cohesion complex, which regulates sister chromatid cohesion and recombination between homologous chromosomes. Multiple alternatively spliced variants, encoding the same protein, have been found for this gene. [provided by RefSeq, Jul 2008].

Specifications

Antigen Phospho-REC8 (Ser251)
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene REC8
Gene Accession No. O95072
Gene Alias Cohesin Rec8p; cohesion rec8p; HR21spB; human homolog of rad21, S. pombe; kleisin-alpha; Mei8; meiotic cohesion Rec8; meiotic recombination and sister chromatid cohesion phosphoprotein of the rad21p family; Meiotic recombination protein REC8 homolog; meiotic recombination protein REC8-like 1; mrec; Rec8; REC8 homolog; REC8 meiotic recombination protein; Rec8L1; REC8-like 1; Rec8p; recombination and sister chromatid cohesion protein homolog
Gene Symbols REC8
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from mouse REC8 around the phosphorylation site of Ser251.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9985
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.