Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Rel (Ser503) Polyclonal Antibody, Abbomax

Catalog No. 502203585
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3585 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3585 Supplier Abbomax Inc Supplier No. 50012024
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: PMA-treated Jurkat cells Cellular location: Nucleus, Cytoplasm.

Phospho-Rel (Ser503) Polyclonal antibody specifically detects Phospho-Rel (Ser503) in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot

Specifications

Antigen Phospho-Rel (Ser503)
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation 25mM tris with proprietary stabilizer and 0.01% sodium azide
Gene REL
Gene Accession No. P15307, Q04864
Gene Alias BCD541; C-Rel; C-Rel protein; C-Rel proto-oncogene protein; gemin 1; gemin-1; LOW QUALITY PROTEIN: proto-oncogene c-Rel; oncogene REL, avian reticuloendotheliosis; proto-oncogene c-Rel; REL; REL proto-oncogene, NF-kB subunit; reticuloendotheliosis oncogene; SMA; SMA1; SMA2; SMA3; SMA4; SMN; SMN2; SMNT; v-rel avian reticuloendotheliosis viral oncogene homolog; v-rel reticuloendotheliosis viral oncogene homolog
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the epitope -TTSSDS-at a phosphorylation site Ser503 of human c-Rel protein
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19696, 5966
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.