Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-REPS1 (Ser272) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5105689
Description
Antibody detects endogenous levels of REPS1 only when phosphorylated at Ser272.
May coordinate the cellular actions of activated EGF receptors and Ral-GTPases. Mungall A.J., Nature 425:805-811(2003). Mural R.J., Submitted (SEP-2005) to the EMBL/GenBank/DDBJ databases. Kimura K., Genome Res. 16:55-65(2006).
Specifications
Phospho-REPS1 (Ser272) | |
Polyclonal | |
Unconjugated | |
REPS1 | |
BB161292; MIG2; RALBP1; RalBP1 associated Eps domain containing 1; RalBP1 associated Eps domain containing protein; ralBP1-associated Eps domain-containing protein 1; RalBP1-interacting protein 1; Reps1 | |
Rabbit | |
sequential chromatography | |
RUO | |
19707, 292944, 85021 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O54916, Q96D71 | |
REPS1 | |
A synthesized peptide derived from human REPS1(Accession Q96D71), corresponding to amino acid residues around phosphorylated Ser272. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction