Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Ret (Tyr1062) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985358
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5358 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5358 Supplier Bioss Supplier No. BS3385R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RET is a member of the cadherin superfamily, encodes one of the receptor tyrosine kinases which are cell-surface molecules that transduce signals for cell growth and differentiation. RET plays a crucial role in neural crest development, can undergo oncogenic activation in vivo and in vitro by cytogenetic rearrangement. Mutations in the RET gene are associated with the disorders multiple endocrine neoplasia, type IIA, multiple endocrine neoplasia, type IIB, Hirschsprung disease, and medullary thyroid carcinoma. Two transcript variants encoding different isoforms have been found for the RET gene. Additional transcript variants have been described for RET but their biological validity has not been confirmed.

Specifications

Antigen Phospho-Ret (Tyr1062)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RET
Gene Accession No. P07949
Gene Alias Cadherin family member 12; cadherin-related family member 16; CDHF12; CDHR16; CG1061; CG14396; CG14396 gene product from transcript CG14396-RA; CG14396-PA; CG14396-PB; CG14396-PC; c-Ret; CUX1/RET fusion; Dmel\CG14396; Dmel_CG14396; DmHD-59; DmRet; DRET; D-ret; EC 2.7.10.1; Extracellular cell-membrane anchored RET cadherin 120 kDa fragment; HD-59; HSCR1; hydroxyaryl-protein kinase; kinase Ret; MEN2; MEN2A; MEN2B; MTC1; OTTHUMP00000216967; Proto-oncogene c-Ret; Proto-oncogene tyrosine-protein kinase receptor Ret; PTC; rearranged during transfection; receptor tyrosine kinase; RET; Ret gene for receptor tyrosin; Ret oncogene; ret proto-oncogene; ret proto-oncogene (multiple endocrine neoplasia and medullary thyroid carcinoma 1, Hirschsprung disease); Ret proto-oncogene (multiple endocrine neoplasia MEN2A MEN2B and medullary thyroid carcinoma 1 Hirschsprung disease); RET receptor tyrosine kinase; RET transforming sequence; RET51; RET9; RET-ELE1; Reto; Ret-PA; Ret-PB; Ret-PC; Soluble RET kinase fragment
Gene Symbols RET
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human Ret around the phosphorylation site of Tyr1062.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5979
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.