Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-RGS19 (Ser151) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118694
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118694 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5118694 Supplier Invitrogen™ Supplier No. PA5118694
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of RGS19 only when phosphorylated at Ser151.

RGS19 inhibits signal transduction by increasing the GTPase activity of G protein alpha subunits thereby driving them into their inactive GDP-bound form. Binds to G-alpha subfamily 1 members, predominantly to G(i)-alpha-3. Activity on G(z)-alpha is inhibited by phosphorylation and palmitoylation of the G-protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-RGS19 (Ser151)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RGS19
Gene Accession No. O70521, P49795, Q9CX84
Gene Alias 2610042F04Rik; AI324841; AW547781; calcium/calmodulin-dependent protein kinase I; Camki; G alpha interacting protein; G protein signalling regulator 19; GAIP; GAIP protein; G-alpha interacting protein; G-alpha-interacting protein; GNAI3IP; GS19; guanine nucleotide binding protein alpha inhibiting activity polypeptide 3 interacting protein; regulator of G-protein signaling 19; regulator of G-protein signalling 19; Rgs19; RGSGAIP
Gene Symbols RGS19
Host Species Rabbit
Immunogen A synthesized peptide derived from human RGS19(Accession P49795), corresponding to amino acid residues around phosphorylated Ser151.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10287, 56470, 59293
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.