Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-RGS19 (Ser151) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986726
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6726 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6726 Supplier Bioss Supplier No. BS5688R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RGS19 inhibits signal transduction by increasing the GTPase activity of G protein alpha subunits thereby driving them into their inactive GDP-bound form. Binds to G-alpha subfamily 1 members, predominantly to G(i)-alpha-3. Activity on G(z)-alpha is inhibited by phosphorylation and palmitoylation of the G-protein.

Specifications

Antigen Phospho-RGS19 (Ser151)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RGS19
Gene Accession No. P49795
Gene Alias 2610042F04Rik; AI324841; AW547781; calcium/calmodulin-dependent protein kinase I; Camki; G alpha interacting protein; G protein signalling regulator 19; Gaip; GAIP protein; G-alpha interacting protein; G-alpha-interacting protein; GNAI3IP; GS19; guanine nucleotide binding protein alpha inhibiting activity polypeptide 3 interacting protein; Regulator of G-protein signaling 19; regulator of G-protein signalling 19; RGS19; RGSGAIP
Gene Symbols RGS19
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human RGS19 around the phosphorylation site of Ser151 [IL (p-S)PK].
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10287
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.