Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-RGS19 (Ser24) Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA512826
Description
Phospho-RGS19 (Ser24) Polyclonal Antibody for DB
RGS19 inhibits signal transduction by increasing the GTPase activity of G protein alpha subunits thereby driving them into their inactive GDP-bound form. Binds to G-alpha subfamily 1 members, predominantly to G(i)-alpha-3. Activity on G(z)-alpha is inhibited by phosphorylation and palmitoylation of the G-protein.
Specifications
Phospho-RGS19 (Ser24) | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
P49795 | |
RGS19 | |
KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S24 of human RGS19. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Dot Blot | |
Unconjugated | |
RGS19 | |
2610042F04Rik; AI324841; AW547781; calcium/calmodulin-dependent protein kinase I; Camki; G alpha interacting protein; G protein signalling regulator 19; GAIP; GAIP protein; G-alpha interacting protein; G-alpha-interacting protein; GNAI3IP; GS19; guanine nucleotide binding protein alpha inhibiting activity polypeptide 3 interacting protein; regulator of G-protein signaling 19; regulator of G-protein signalling 19; Rgs19; RGSGAIP | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
10287 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction