Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-RhoGDI (Ser174) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538827
Description
Aplysia Ras-related homologs (ARHs), also called Rho genes, belong to the RAS gene superfamily encoding small guanine nucleotide exchange (GTP/GDP) factors. The ARH proteins may be kept in the inactive, GDP-bound state by interaction with GDP dissociation inhibitors, such as ARHGDIA (Leffers et al., 1993 [PubMed 8262133]).
Specifications
| Phospho-RhoGDI (Ser174) | |
| Polyclonal | |
| Unconjugated | |
| ARHGDIA | |
| Rho GDP-dissociation inhibitor 1, Rho GDI 1, GDI-1, Rho-GDI alpha, Arhgdia, C87222, Gdi1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 192662, 360678, 396 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P52565, Q99PT1, Q5XI73 | |
| ARHGDIA | |
| A synthetic phosphopeptide derived from human ARHGDIA around the phosphorylation site of Ser174 (R-G-SP-Y-S) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction