Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-RICTOR (Thr1135) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104644
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104644 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104644 Supplier Invitrogen™ Supplier No. PA5104644
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of Rictor only when phosphorylated at Thr1135.

RICTOR and MTOR (FRAP1) are components of a protein complex that integrates nutrient- and growth factor-derived signals to regulate cell growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-RICTOR (Thr1135)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Rictor
Gene Accession No. Q6QI06, Q6R327
Gene Alias 4921505C17Rik; 6030405M08Rik; AVO3; AVO3 homolog; AW492497; D530039E11Rik; DKFZp686B11164; Fyb; FYN binding protein; hAVO3; Kiaa1999; mAVO3; MGC39830; PIA; pianissimo; protein pianissimo; Rapamycin-insensitive companion of mTOR; rictor; RPTOR independent companion of MTOR complex 2; RPTOR independent companion of MTOR, complex 2; TORC2-specific protein AVO3
Gene Symbols Rictor
Host Species Rabbit
Immunogen A synthesized peptide derived from human RICTOR(Accession Q6R327), corresponding to amino acid residues around phosphorylated Thr1135.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 253260, 78757
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.