Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-RIP1 (Ser166) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5117250

Description
Positive Samples: NIH/3T3
Phospho-RIP1 (Ser166) Polyclonal antibody specifically detects Phospho-RIP1 (Ser166) in Human, Mouse, Mouse samples. It is validated for Western Blot
Specifications
Phospho-RIP1 (Ser166) | |
Polyclonal | |
Unconjugated | |
RIPK1 | |
Cell death protein RIP; D330015H01Rik; FLJ39204; OTTHUMP00000015955; receptor (TNFRSF)-interacting serine-threonine kinase 1; receptor interacting serine/threonine kinase 1; receptor-interacting protein 1; receptor-interacting protein kinase 1; receptor-interacting serine/threonine-protein kinase 1; Rinp; RIP; RIP1; RIP-1; RIP1 (CT); RIPK1; serine/threonine-protein kinase RIP | |
A phospho specific peptide corresponding to residues surrounding S166 of Mouse RIPK1 | |
100 μL | |
Primary | |
Human, Mouse, Mouse | |
Antibody | |
IgG |
Western Blot | |
3.01 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13546, Q60855 | |
Rabbit | |
Affinity chromatography | |
RUO | |
19766, 8737 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction