Learn More
Phospho-RIPK1 (Ser161) Mouse anti-Human, Unconjugated, Clone: 1B2G1, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | Phospho-RIPK1 (Ser161) |
|---|---|
| Clone | 1B2G1 |
| Concentration | 1468 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-223-9053
![]()
|
Proteintech Group Inc
668541IG100UL |
100 μL |
Each of 1 for $518.60
|
|
|||||
Description
The Phospho-RIPK1 (Ser161) antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human Phospho-RIPK1 (Ser161) . This antibody recognizes human antigen. The Phospho-RIPK1 (Ser161) antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| Phospho-RIPK1 (Ser161) | |
| 1468 μg/mL | |
| Monoclonal | |
| Mouse | |
| 8737 | |
| Primary |
| 1B2G1 | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Human | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.