Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-RNA pol II CTD (Ser2) Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501992721
Encompass
Change view
Click to view available options
Quantity:
10 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2721 200 μL
50-199-2722 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2721 Supplier Active Motif Supplier No. 39563
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RNA polymerase II is the enzyme responsible for the synthesis of mRNA and many non-coding RNAs. POLR2A encodes RNA polymerase II subunit B1 (RPB1), the largest subunit of RNA polymerase II. The C-terminal domain (CTD) of RPB1 contains tandem heptapeptide repeats with the consensus sequence YSPTSPS. Phosphorylation and other modifications within this sequence influence the recruitment of factors that regulate the transcription cycle. Recruitment of RNA processing factors to the elongation complex is coordinated by the phosphorylation of Serine-2 and Serine-5 residues within the CTD.

Specifications

Antigen Phospho-RNA pol II CTD (Ser2)
Applications Dot Blot, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene POLR2A
Gene Accession No. P09884, P24928
Gene Alias 220kDa; AW321876; DNA polymerase alpha 1 catalytic subunit; DNA polymerase alpha 1, 180 kDa; DNA polymerase alpha 1, catalytic subunit; DNA polymerase alpha catalytic subunit; DNA polymerase alpha catalytic subunit p180; DNA polymerase alpha p180 subunit; DNA polymerase alpha subunit I; DNA polymerase epsilon catalytic subunit; DNA polymerase epsilon catalytic subunit A; DNA polymerase II subunit A; DNA-directed RNA polymerase II largest subunit, RNA polymerase II 220 kd subunit; DNA-directed RNA polymerase II subunit A; DNA-directed RNA polymerase II subunit RPB1; DNA-directed RNA polymerase III largest subunit; DUN2; EC 2.7.7.6; G7a/Bat6Hom; hRPB220; hsRPB1; I79_002751; I79_016081; MGC75453; N0825; NSX; p180; Pol II S2p; POL2; Pola; POLA1; POLR2; Polr2a; POLRA; polymerase (DNA directed), alpha 1; polymerase (DNA directed), alpha 1, catalytic subunit; polymerase (DNA) alpha 1, catalytic subunit; polymerase (DNA-directed), alpha (70kD); Polymerase (RNA II (DNA directed), large polypeptide; polymerase (RNA) II (DNA directed) polypeptide A; polymerase (RNA) II (DNA directed) polypeptide A, 220kDa; polymerase (RNA) II subunit A; POLYMERASE II; polymerase, alpha 1; RNA; RNA POL2; RNA polymerase II 1; RNA polymerase II largest subunit; RNA polymerase II subunit A; RNA polymerase II subunit B1; RNA-directed RNA polymerase II subunit RPB1; RP02; RPB1; RPB220; RPBh1; Rpii215; RpIILS; RPO2; RPO21; Rpo2-1; RPOL2; SUA8; YNL262W
Gene Symbols POLA1, POLR2A
Host Species Rabbit
Immunogen This RNA pol II CTD phospho Ser2 antibody was raised against a peptide containing the RNA Pol II CTD sequence phosphorylated at serine 2.
Purification Method Antigen affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5422, 5430
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.