Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-RNA pol II CTD (Ser5) Recombinant Mouse Monoclonal Antibody, Active Motif™

Mouse Recombinant Monoclonal Antibody
$110.70 - $492.00
Specifications
| Antigen | Phospho-RNA pol II CTD (Ser5) |
|---|---|
| Concentration | 1 mg/mL |
| Applications | ChIP sequencing (ChIP-seq), ELISA, Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3712
![]()
|
Active Motif
91120 |
10 μg |
Each for $110.70
|
|
|||||
50-199-3711
![]()
|
Active Motif
91119 |
100 μg |
Each for $492.00
|
|
|||||
Description
RNA polymerase II is the enzyme responsible for the synthesis of mRNA and many non-coding RNAs. POLR2A encodes RNA polymerase II subunit B1 (RPB1), the largest subunit of RNA polymerase II. The C-terminal domain (CTD) of RPB1 contains tandem heptapeptide repeats with the consensus sequence YSPTSPS. Phosphorylation and other modifications within this sequence influence the recruitment of factors that regulate the transcription cycle. Recruitment of RNA processing factors to the elongation complex is coordinated by the phosphorylation of Serine-2 and Serine-5 residues within the CTD.Specifications
| Phospho-RNA pol II CTD (Ser5) | |
| ChIP sequencing (ChIP-seq), ELISA, Immunocytochemistry, Immunofluorescence, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P09884, P24928 | |
| 5422, 5430 | |
| This RNA pol II CTD phospho Ser5 antibody was raised against a peptide containing the RNA pol II CTD sequence phosphorylated at serine 5. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| POLR2A |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 200mM HEPES, 50mM sodium acetate with 100mM NaCl, 30% glycerol and 0.035% sodium azide; pH 7.5 | |
| 220kDa; AW321876; DNA polymerase alpha 1 catalytic subunit; DNA polymerase alpha 1, 180 kDa; DNA polymerase alpha 1, catalytic subunit; DNA polymerase alpha catalytic subunit; DNA polymerase alpha catalytic subunit p180; DNA polymerase alpha p180 subunit; DNA polymerase alpha subunit I; DNA polymerase epsilon catalytic subunit; DNA polymerase epsilon catalytic subunit A; DNA polymerase II subunit A; DNA-directed RNA polymerase II largest subunit, RNA polymerase II 220 kd subunit; DNA-directed RNA polymerase II subunit A; DNA-directed RNA polymerase II subunit RPB1; DNA-directed RNA polymerase III largest subunit; DUN2; EC 2.7.7.6; G7a/Bat6Hom; hRPB220; hsRPB1; I79_002751; I79_016081; MGC75453; N0825; NSX; p180; Pol II S2p; POL2; Pola; POLA1; POLR2; Polr2a; POLRA; polymerase (DNA directed), alpha 1; polymerase (DNA directed), alpha 1, catalytic subunit; polymerase (DNA) alpha 1, catalytic subunit; polymerase (DNA-directed), alpha (70kD); Polymerase (RNA II (DNA directed), large polypept | |
| POLA1, POLR2A | |
| IgG2a | |
| Metal-chelate chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title