Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-RPA2 (Ser4, Ser8) Recombinant Rabbit Monoclonal Antibody (BL-165-5F1)
Rabbit Recombinant Monoclonal Antibody
$173.10 - $436.00
Specifications
Antigen | Phospho-RPA2 (Ser4, Ser8) |
---|---|
Clone | BL-165-5F1 |
Concentration | 1 mg/mL |
Content And Storage | 4°C |
Applications | Immunoprecipitation, Western Blot, Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA544213
|
Invitrogen™
MA544213 |
100 μL |
Each for $436.00
|
|
|||||
PIMA544214
|
Invitrogen™
MA544214 |
20 μL |
Each for $173.10
|
|
|||||
Description
Phospho-RPA2 (Ser4, Ser8) Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IP
Required for DNA recombination, repair and replication. The activity of RP-A is mediated by single-stranded DNA binding and protein interactions. Required for the efficient recruitment of the DNA double-strand break repair factor RAD51 to chromatin in response to DNA damage.Specifications
Phospho-RPA2 (Ser4, Ser8) | |
1 mg/mL | |
Immunoprecipitation, Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
BBS with 0.09% sodium azide; pH 8.2 | |
P15927 | |
6118 | |
Dually phosphorylated synthetic peptide, which represented a portion of human replication protein A2, 32 kDa surrounding phosphorylated serines that corresponded to positions 4 and 8. | |
Antibody |
BL-165-5F1 | |
4°C | |
Recombinant Monoclonal | |
Liquid | |
IgG | |
Human | |
Rpa2 | |
30-kDa protein; AA409079; AI325195; AU020965; HSSB; hypothetical protein; I79_004200; p32; p32-subunit of replication protein A; p34; REPA2; Replication factor A protein 2; replication protein A 32 kDa subunit; Replication protein A 32kDa subunit; Replication protein A 34 kDa subunit; Replication protein A 34kDa subunit; replication protein A2; replication protein A2, 32kDa; replication protein A4; RFA; RF-A protein 2; Rf-A2; RP21C; RP4-547C9.3; RP-A p32; RP-A p34; RPA2; RPA32; RPA34; RPA4; RpLP1; RpP2 | |
Rpa2 | |
Primary | |
purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title