Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-RPA2Thr21 (Thr21) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986727
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6727 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6727 Supplier Bioss Supplier No. BS5693R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Required for DNA recombination, repair and replication. The activity of RP-A is mediated by single-stranded DNA binding and protein interactions. Required for the efficient recruitment of the DNA double-strand break repair factor RAD51 to chromatin in response to DNA damage.

Specifications

Antigen Phospho-RPA2Thr21 (Thr21)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Rpa2
Gene Accession No. P15927
Gene Alias 30-kDa protein; AA409079; AI325195; AU020965; HSSB; p32-subunit of replication protein A; RCJMB04_6d17; REPA2; Replication factor A protein 2; Replication protein A 32 kDa subunit; Replication protein A 34 kDa subunit; replication protein A2; replication protein A2, 32kDa; RF-A protein 2; Rf-A2; RP4-547C9.3; RP-A p32; RP-A p34; Rpa2; RPA32; RPA34
Gene Symbols Rpa2
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human RPA2 around the phosphorylation site of Thr21.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6118
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.