Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-RPA32 (Ser4, Ser8) Recombinant Rabbit Monoclonal Antibody (BL-165-5F1), Bethyl Laboratories

Catalog No. 501571272
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
20 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-1272 20 μL
50-157-1271 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-1272 Supplier Bethyl Laboratories, Inc Supplier No. A700009T
Add to Cart
Add to Cart

Rabbit Recombit Monoclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Required for DNA recombination, repair and replication. The activity of RP-A is mediated by single-stranded DNA binding and protein interactions. Required for the efficient recruitment of the DNA double-strand break repair factor RAD51 to chromatin in response to DNA damage.

Specifications

Antigen Phospho-RPA32 (Ser4,Ser8)
Applications Immunoprecipitation, Western Blot
Classification Recombinant Monoclonal
Clone BL-165-5F1
Conjugate Unconjugated
Formulation BBS with 0.09% sodium azide; pH 8.2
Gene RPA2
Gene Accession No. P15927
Gene Alias REPA2, RPA32
Gene Symbols RPA2
Host Species Rabbit
Immunogen Surrounding serine 4/8
Purification Method Ion-exchange chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6118
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.