Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-RSK Pan (Ser221, Ser227, Ser218, Ser232) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114637
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114637 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114637 Supplier Invitrogen™ Supplier No. PA5114637
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of RSK1/2/3/4 only when phosphorylated at Ser221/227/s218/232.

The p90 ribosomal S6 kinases (RSKs) comprise a family of serine/threonine kinases that lie at the terminus of the ERK pathway. In humans, the RSK family consists of four isoforms (RSK1 to-4). RSK family members are unusual among serine/threonine kinases in that they contain two distinct kinase domains, both of which are catalytically functional. Theses kinase domains are activated in a sequential manner by a series of phosphorylations. RSK regulates gene expression via association and phosphorylation of transcriptional regulators including c-Fos, estrogen receptor, NFkappaB/IkappaB alpha, cAMP-response element-binding protein (CREB). ERK activates the C-terminal kinase of RSK, leading to activation of the N-terminal kinase. Members of the RSK family are present in the cytoplasm as well as the nucleus. Addition of growth factor to the cells results in the activation of both cytosolic and nuclear RSK and the translocation of the cytosolic RSK into the nucleus upon activation. The activation and nuclear translocation of RSK result in phosphorylation and activation of transcription factors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-RSK Pan (Ser221, Ser227, Ser218, Ser232)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene RPS6KA1
Gene Accession No. O75676, P18653, P18654, P51812, Q15349, Q15418, Q63531, Q7TPS0, Q9UK32, Q9WUT3, Q9Z2B9
Gene Alias 1110069D02Rik; 2610524K04Rik; 90 kDa ribosomal protein S6 kinase 1; 90 kDa ribosomal protein S6 kinase 2; 90 kDa ribosomal protein S6 kinase 3; 90 kDa ribosomal protein S6 kinase 4; 90 kDa ribosomal protein S6 kinase 6; 90kDa; AI848992; C130006E23; CLS; D17Wsu134e; dJ590P13.1 (ribosomal protein S6 kinase, 90kD, polypeptide 1); HU-1; HU-2; HU-3; Insulin-stimulated protein kinase 1; ISPK1; ISPK-1; MAP kinase-activated protein kinase 1a; MAP kinase-activated protein kinase 1b; MAP kinase-activated protein kinase 1c; MAPK-activated protein kinase 1a; MAPK-activated protein kinase 1b; MAPK-activated protein kinase 1c; MAPKAP kinase 1a; MAPKAP kinase 1b; MAPKAP kinase 1c; MAPKAPK1A; MAPKAPK-1a; MAPKAPK1B; MAPKAPK-1b; MAPKAPK1C; MAPKAPK-1c; MAPKAPK1D; mitogen- and stress-activated protein kinase 2; mitogen- and stress-activated protein kinase-2; mitogen-activated protein kinase-activated protein kinase 1C; mMSK2; MPK-9; MRX19; Msk2; Nuclear mitogen- and stress-activated protein kinase 2; p90 rsk; p90rsk; p90-RSK 1; p90-RSK 2; p90-RSK 3; p90-RSK 6; p90RSK1; p90-Rsk1; p90RSK2; p90-RSK2; p90RSK3; p90-RSK3; p90RSK6; p90S6K; pp90rsk; pp90RSK2; pp90RSK3; PP90RSK4; Protein-tyrosine kinase Mpk-9; ribosomal protein kinase B; ribosomal protein S6 kinase 2; ribosomal protein S6 kinase A1; ribosomal protein S6 kinase A2; ribosomal protein S6 kinase A3; ribosomal protein S6 kinase A4; ribosomal protein S6 kinase A6; ribosomal protein S6 kinase alpha 1; ribosomal protein S6 kinase alpha 2; ribosomal protein S6 kinase alpha 4; ribosomal protein S6 kinase alpha-1; Ribosomal protein S6 kinase alpha-2; ribosomal protein S6 kinase alpha-3; Ribosomal protein S6 kinase alpha-4; ribosomal protein S6 kinase alpha-6; ribosomal protein S6 kinase polypeptide 1; ribosomal protein S6 kinase polypeptide 3; ribosomal protein S6 kinase polypeptide 6; ribosomal protein S6 kinase, 90kD, polypeptide 1; Ribosomal protein S6 kinase, 90kD, 1; ribosomal protein S6 kinase, 90kD, polypeptide 2; ribosomal protein S6 kinase, 90kD, polypeptide 4; ribosomal protein S6 kinase, 90kDa, polypeptide 1; ribosomal protein S6 kinase, 90kDa, polypeptide 2; ribosomal protein S6 kinase, 90kDa, polypeptide 3; ribosomal protein S6 kinase, 90kDa, polypeptide 4; ribosomal protein S6 kinase, 90kDa, polypeptide 6; ribosomal protein S6 kinase, polypeptide 2; ribosomal protein S6 kinase, polypeptide 4; ribosomal S6 kinase 1; Ribosomal S6 kinase 2; ribosomal S6 kinase 3; Ribosomal S6 kinase 4; RLSK; RP11-393H10.3; Rps6ka1; RPS6KA2; Rps6ka3; RPS6KA4; RPS6KA6; Rps6ka-rs1; RSK; Rsk1; RSK-1; Rsk2; RSK-2; Rsk3; RSK-3; RSK4; RSK-4; RSKB; RSK-B; RSK-like protein kinase; S6K-alpha; S6K-alpha 1; S6K-alpha 2; S6K-alpha 6; S6K-alpha-1; S6K-alpha2; S6K-alpha-2; S6K-alpha3; S6K-alpha-3; S6K-alpha-4; S6K-alpha-6
Gene Symbols RPS6KA1, RPS6KA2, Rps6ka3, RPS6KA4, RPS6KA6
Host Species Rabbit
Immunogen A synthesized peptide derived from human RSK1/2/3/4 around the phosphorylation site of Ser221/227/s218/232.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110651, 117269, 20111, 20112, 27330, 317203, 361715, 501560, 56613, 6195, 6196, 6197, 67071, 81771, 8986
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.