Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-RSK1 (Ser352) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA537729
Description
A suggested positive control for Western blot is HUVEC cells.
RPS6KA1 (RSK1) is a growth-factor regulated serine/threonine kinase that is involved in the MAPK cascade. It is thought to control cellular proliferation and differentiation through the phosphorylation of transcription factors. RPS6KA1 contains 2 nonidentical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
Phospho-RSK1 (Ser352) | |
Polyclonal | |
Unconjugated | |
RPS6KA1 | |
90 kDa ribosomal protein S6 kinase 1; dJ590P13.1 (ribosomal protein S6 kinase, 90kD, polypeptide 1); hu-1; MAP kinase-activated protein kinase 1; MAP kinase-activated protein kinase 1a; MAPK-activated protein kinase 1; MAPK-activated protein kinase 1a; MAPKAP kinase 1; MAPKAP kinase 1a; MAPKAPK-1; mapkapk1a; MAPKAPK-1a; p90Rsk; P90-RSK; p90-RSK 1; p90Rsk1; p90-Rsk1; p90S6K; pp90RSK1; ribosomal protein S6 kinase 2 alpha; ribosomal protein S6 kinase 2 beta; ribosomal protein S6 kinase a, polypeptide 1; ribosomal protein S6 kinase A1; ribosomal protein S6 kinase A1 L homeolog; ribosomal protein S6 kinase alpha 1; Ribosomal protein S6 kinase alpha-1; Ribosomal protein S6 kinase II alpha; ribosomal protein S6 kinase II beta; ribosomal protein S6 kinase polypeptide 1; ribosomal protein S6 kinase, 90kD, polypeptide 1; Ribosomal protein S6 kinase, 90kD, 1; ribosomal protein S6 kinase, 90kDa, polypeptide 1; ribosomal protein S6 kinase, 90kDa, polypeptide 1 L homeolog; ribosomal protein S6 kinase, polypeptide A1; ribosomal S6 kinase 1; RPS6KA; Rps6ka1; rps6ka1.L; RPS6KA1L; rsk; RSK1; Rsk-1; S6 kinase II beta; S6 protein kinase (Rsk-1); S6K-alpha 1; S6K-alpha-1; S6KII-alpha; S6KII-beta; wu:fc56h07; XELAEV_18012109mg; zgc:152654 | |
Rabbit | |
Affinity chromatography | |
RUO | |
20111, 6195, 81771 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P18653, Q15418, Q63531 | |
RPS6KA1 | |
Peptide sequence around phosphorylation site of serine 352 (R-D-S(p)-P-G) derived from Human p90RSK. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction