Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-RSK2 (Tyr529) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101034
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101034 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101034 Supplier Invitrogen™ Supplier No. PA5101034
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of RSK2 only when phosphorylated at Tyr529.

RPS6KA3 (RSK2) is a serine/threonine kinase that modulates many cellular processes including transcription, adhesion, growth, survival, and proliferation. RPS6KA3 contains 2 non-identical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Mutations in the gene have been associated with Coffin-Lowry syndrome (CLS).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-RSK2 (Tyr529)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RPS6KA3
Gene Accession No. P18654, P51812
Gene Alias 90 kDa ribosomal protein S6 kinase 3; C130006E23; CLS; HU-3; Insulin-stimulated protein kinase 1; ISPK1; ISPK-1; MAP kinase-activated protein kinase 1b; MAPK-activated protein kinase 1b; MAPKAP kinase 1b; MAPKAPK1B; MAPKAPK-1b; MPK-9; MRX19; p90-RSK 3; p90-RSK2; p90RSK3; pp90RSK2; RGD1563860; ribosomal protein S6 kinase 2; ribosomal protein S6 kinase A3; ribosomal protein S6 kinase alpha-3; ribosomal protein S6 kinase polypeptide 3; ribosomal protein S6 kinase, 90kDa, polypeptide 3; ribosomal S6 kinase 2; Rps6ka3; Rps6ka-rs1; RSK; Rsk2; RSK-2; S6K-alpha3; S6K-alpha-3
Gene Symbols RPS6KA3
Host Species Rabbit
Immunogen A synthesized peptide derived from human RPS6KA3(Accession P51812), corresponding to amino acid residues around phosphorylated Tyr529.
Purification Method Sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110651, 6197
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.