Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-RSK3 (Thr356, Ser360) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585817
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585817 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585817 Supplier Invitrogen™ Supplier No. PA585817
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (100%), Rat (100%), Bovine (100%). Positive Control: EGF treated A431. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

RPS6KA2 (RSK3) serine/threonine kinase is a member of the ribosomal S6 kinase (RSK) family. It is believed that RPS6KA2 is a signal-transducing intermediate in the cellular response to several growth factors. It is found in many tissues, but in higher levels in lung and skeletal muscle. The p90 ribosomal S6 kinases (RSK)1-4 are downstream members of the extracellular signal-regulated kinase (ERK)/MAPK cascade. The loss of RSK2 activity in humans leads to Coffin-Lowry syndrome, which is characterized by mental retardation and growth deficits and may have a tonic regulation on G-protein coupled signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-RSK3 (Thr356, Ser360)
Applications Western Blot
Classification Polyclonal
Concentration 0.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene RPS6KA2
Gene Accession No. Q15349
Gene Alias 90 kDa ribosomal protein S6 kinase 2; 90kDa; D17Wsu134e; HU-2; MAP kinase-activated protein kinase 1c; MAPK-activated protein kinase 1c; MAPKAP kinase 1C; MAPKAPK1C; MAPKAPK-1c; mitogen-activated protein kinase-activated protein kinase 1C; p90rsk; p90-RSK 2; p90RSK2; p90-RSK3; pp90rsk; pp90RSK3; Protein-tyrosine kinase Mpk-9; ribosomal protein S6 kinase A2; ribosomal protein S6 kinase alpha 2; ribosomal protein S6 kinase alpha-2; ribosomal protein S6 kinase polypeptide 2; ribosomal protein S6 kinase, 90kD, polypeptide 2; ribosomal protein S6 kinase, 90kDa, polypeptide 2; ribosomal protein S6 kinase, polypeptide 2; ribosomal S6 kinase 3; Rps6ka2; Rps6ka-rs1; RSK; Rsk3; RSK-3; S6K-alpha; S6K-alpha 2; S6K-alpha2; S6K-alpha-2
Gene Symbols RPS6KA2
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide surrounding phospho Thr356/Ser360 of human RSK3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6196
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.