Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phospho-SAMHD1 (Thr592) Mouse anti-Human, Unconjugated, Clone: , MilliporeSigma™

Catalog No. MABF93425UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF934-25UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF934-25UL Supplier MilliporeSigma Supplier No. MABF93425UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-phospho-SAMHD1 (Thr592), clone T592P, Cat. No. MABF934 is a mouse monoclonal antibody that detects SAMHD1 and has been tested for use in Flow Cytometry and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-SAMHD1 (Thr592)
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Deoxynucleoside triphosphate triphosphohydrolase SAMHD1;dNTPase;Dendritic cell-derived IFNG-induced protein;DCIP;Monocyte protein 5;MOP-5;SAM domain and HD domain-containing protein 1
Gene Symbols SAMHD1;MOP5
Host Species Mouse
Immunogen A linear peptide corresponding to 11 amino acids surrounding phosphoryalted Thr592 from the C-terminal region of huamn SAMHD1.
Molecular Weight of Antigen ∽72 kDa observed; 72.20 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Test Specificity Clone T592P detects Deoxynucleoside triphosphate triphosphohydrolase SAMHD1 in human cells. It targets an epitope with in 11 amino acids surrounding the phosphorylated Thr592 from the C-terminal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.