Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-SAPK3 (Thr183, Tyr185) Rabbit anti-Human∣Mouse∣Rat, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS12001R

Description
Activation of members of the mitogen-activated protein kinase family is a major mechanism for transduction of extracellular signals. Stress-activated protein kinases are one subclass of MAP kinases. MAPK12 functions as a signal transducer during differentiation of myoblasts to myotubes.Specifications
Phospho-SAPK3 (Thr183, Tyr185) | |
Polyclonal | |
Unconjugated | |
Mapk12 | |
AW123708; ERK3; ERK5; ERK6; ERK-6; etID309866.18; Extracellular signal-regulated kinase 6; fa05c12; MAP kinase 12; MAP kinase p38 gamma; MAPK 12; MAPK12; mapk12a; mitogen activated protein kinase 12; Mitogen-activated protein kinase 12; mitogen-activated protein kinase 12a; mitogen-activated protein kinase 3; Mitogen-activated protein kinase p38 gamma; OTTHUMP00000196671; P38GAMMA; p38-gamma; Prkm12; SAP kinase-3; SAPK3; SAPK-3; stress activated protein kinase 3; Stress-activated protein kinase 3; stress-activated protein kinase-3; wu:fa05c12; zgc:101695 | |
Rabbit | |
Protein A | |
RUO | |
29857, 60352, 6300 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
O08911, P53778, Q63538 | |
Mapk12 | |
KLH conjugated synthetic phosphopeptide derived from human SAPK3 around the phosphorylation site of Thr183+Tyr185 [EM (p-T)G (p-Y)VV]. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction