Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-MAP2K4 (Ser80) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104908
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104908 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104908 Supplier Invitrogen™ Supplier No. PA5104908
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of MEK4/MKK4 only when phosphorylated at Serine 80.

MAP2K4 (MEK4) serine/threonine kinase (mitogen-activated protein kinase 4) MEK4 or MKK4 is a dual specificity kinase of the STE7 family that phosphorylates and activates JNK1 and -2 as well as p38. MAP2K4 preferentially phosphorylates the tyrosine residue of JNKs. MAP2K4 mediates a variety of physiological and pathophysiological processes such as responses to cellular stresses and inflammatory cytokines. MAP2K4 is important for carcinogenesis; a loss-off-function mutation in MAP2K4 was found in lung and pancreatic tumors. Conversely, a pro-oncogenic role was also found for MAP2K4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-MAP2K4 (Ser80)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MAP2K4
Gene Accession No. P45985, P47809
Gene Alias C-JUN N-terminal kinase kinase 1; dual specificity mitogen-activated protein kinase kinase 2; dual specificity mitogen-activated protein kinase kinase 4; dual specificity mitogen-activated protein kinase kinase 4; dual specificity mitogen-activated protein kinase kinase 4a; JNK kinase 1; JNK-activated kinase 1; JNK-activating kinase 1; JNKK; JNKK 1; JNKK1; MAP kinase activator 2; MAP kinase kinase 4; Map2k4; map2k4.L; map2k4a; map2k4-a; map2k4-b; MAP2K4delta; MAPK/ERK kinase 4; MAPKK 4; MAPKK4; MEK 4; MEK4; mitogen activated protein kinase kinase 4; mitogen activated protein kinase kinase 4, variant 1; mitogen-activated protein kinase kinase 4; mitogen-activated protein kinase kinase 4 L homeolog; mitogen-activated protein kinase kinase 4a; MKK4; mkk4a; MKK4A-l; PRKMK4; SAPK kinase 1; SAPK/ERK kinase 1; SAPK/Erk/kinase 1; SAPKK1; SAPKK-1; Sek1; Serk1; Skk1; Stress-activated protein kinase kinase 1; XELAEV_18044157mg; XMEK2; zgc:136500; zMKK4 protein
Gene Symbols MAP2K4
Host Species Rabbit
Immunogen A synthesized peptide derived from human MAP2K4(Accession P45985), corresponding to amino acid residues around phosphorylated Ser80.
Purification Method Sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26398, 287398, 6416
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.