Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-Separase (Ser1126) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540189
Description
Phospho-Separase (Ser1126) Polyclonal Antibody for Western Blot, ICC/IF
Caspase-like protease, which plays a central role in the chromosome segregation by cleaving the SCC1/RAD21 subunit of the cohesin complex at the onset of anaphase. During most of the cell cycle, it is inactivated by different mechanisms.
Specifications
Phospho-Separase (Ser1126) | |
Polyclonal | |
Unconjugated | |
ESPL1 | |
AL024103; AU045071; Caspase-like protein ESPL1; Cerp; Cut1/ESP1 related protein; ESP1; ESPL1; extra spindle pole bodies 1 (S. cerevisiae); extra spindle pole bodies 1, separase; extra spindle pole bodies homolog 1; extra spindle pole bodies homolog 1 (S. cerevisiae); extra spindle pole bodies like 1, separase; extra spindle poles like 1; extra spindle poles-like 1 protein; FLJ46492; KIAA0165; PRCE; SEPA; separase; Separin; separin, cysteine protease; SSE | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9700 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q14674 | |
ESPL1 | |
Synthesized peptide derived from human ESPL1 around phosphorylation site of S1126. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction