Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Separase (Ser801) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538245
Description
Stable cohesion between sister chromatids before anaphase and their timely separation during anaphase are critical for chromosome inheritance. In vertebrates, sister chromatid cohesion is released in 2 steps via distinct mechanisms. The first step involves phosphorylation of STAG1 or STAG2 in the cohesin complex. The second step involves cleavage of the cohesin subunit SCC1 by ESPL1, or separase, which initiates the final separation of sister chromatids .
Specifications
| Phospho-Separase (Ser801) | |
| Polyclonal | |
| Unconjugated | |
| ESPL1 | |
| ESP1, FLJ46492, KIAA0165, caspase-like protein ESPL1, extra spindle poles like 1, extra spindle poles-like 1 protein, separin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 105988, 9700 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q14674, P60330 | |
| ESPL1 | |
| A synthetic phosphopeptide derived from human SEPARASE around the phosphorylation site of Ser801 (N-S-SP-P-V) | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction