Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-SF3B1 (Thr313) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104648
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104648 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104648 Supplier Invitrogen™ Supplier No. PA5104648
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of SF3B1 only when phosphorylated at Thr313.

This gene encodes subunit 1 of the splicing factor 3b protein complex. Splicing factor 3b, together with splicing factor 3a and a 12S RNA unit, forms the U2 small nuclear ribonucleoproteins complex. The splicing factor 3b/3a complex binds pre-mRNA upstream of the intron's branch site in a sequence independent manner and may anchor the U2 snRNP to the pre-mRNA. Splicing factor 3b is also a component of the minor U12-type spliceosome. The carboxy-terminal two-thirds of subunit 1 have 22 non-identical, tandem HEAT repeats that form rod-like, helical structures. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-SF3B1 (Thr313)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SF3B1
Gene Accession No. O75533, Q99NB9
Gene Alias 155kDa; 2810001M05Rik; AA409119; Hsh155; LOW QUALITY PROTEIN: splicing factor 3B subunit 1; MDS; pre-mRNA processing 10; pre-mRNA splicing factor SF3b, 155 kDa subunit; pre-mRNA-splicing factor SF3b 155 kDa subunit; PRP10; PRPF10; SAP 155; SAP155; SF3B1; SF3b155; spliceosome-associated protein 155; Splicing factor 3B subunit 1; splicing factor 3b subunit 1, 155kDa; splicing factor 3b, subunit 1; splicing factor 3b, subunit 1, 155 kDa; splicing factor 3b, subunit 1, 155kD; splicing factor 3b, subunit 1, 155kDa; TA-8; Targ4; transforming growth factor alpha regulated gene 4; tst; wu:fb99f09
Gene Symbols SF3B1
Host Species Rabbit
Immunogen A synthesized peptide derived from human SF3B1(Accession O75533), corresponding to amino acid residues around phosphorylated Thr313.
Purification Method sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23451, 81898, 84486
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.