Learn More
Invitrogen™ Phospho-SGK223 (Tyr413) Polyclonal Antibody

Description
This antibody is predicted to react with mouse and rat based on sequence homology.
Specifications
Specifications
| Antigen | Phospho-SGK223 (Tyr413) |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | PRAG1 |
| Gene Accession No. | Q571I4, Q86YV5 |
| Gene Alias | 9830148H23; D8Ertd82e; DNA segment, Chr 8, ERATO Doi 82, expressed; homolog of rat pragma of Rnd2; Inactive tyrosine-protein kinase PRAG1; mFLJ00269; NACK; Notch activation complex kinase; PEAK1-related kinase-activating pseudokinase 1; Prag1; Pragma of Rnd2; pragmin; RGD1311793; Sgk223; Sugen kinase 223; tyrosine-protein kinase SgK223 |
| Gene Symbols | PRAG1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.