Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-SGK3 (Thr320) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104649
Description
Antibody detects endogenous levels of SGK3 only when phosphorylated at T320.
SGKL (SGK3) is a serine/threonine protein kinase which is similar to serum- and glucocorticoid-induced protein kinase (SGK). Unlike SGK, SGKL is not induced by serum or glucocorticoids. It is induced in response to signals that activate phosphatidylinositol 3-kinase, which is also true for SGK. The protein phosphorylates several target proteins and has a role in neutral amino acid transport and activation of potassium and chloride channels. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
Phospho-SGK3 (Thr320) | |
Polyclonal | |
Unconjugated | |
SGK3 | |
2510015P22Rik; A330005P07Rik; CISK; cytokine-independent survival kinase; DKFZp781N0293; fy; fz; Serine/threonine-protein kinase Sgk3; serum glucocorticoid regulated kinase 3; serum/glucocorticoid regulated kinase 3; serum/glucocorticoid regulated kinase family member 3; serum/glucocorticoid regulated kinase family, member 3; serum/glucocorticoid-regulated kinase 3; Serum/glucocorticoid-regulated kinase-like; SGK2; SGK3; Sgkl | |
Rabbit | |
Sequential Chromatography | |
RUO | |
170755, 171498, 23678 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8R4V0, Q96BR1, Q9ERE3 | |
SGK3 | |
A synthesized peptide derived from human SGK3(Accession Q96BR1), corresponding to amino acid residues around phosphorylated Thr320. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction