Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-SGOL1 (Ser14) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118715
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118715 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5118715 Supplier Invitrogen™ Supplier No. PA5118715
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of SGOL1 only when phosphorylated at Ser14.

The protein encoded by this gene is a member of the shugoshin family of proteins. This protein is thought to protect centromeric cohesin from cleavage during mitotic prophase by preventing phosphorylation of a cohesin subunit. Reduced expression of this gene leads to the premature loss of centromeric cohesion, mis-segregation of sister chromatids, and mitotic arrest. Evidence suggests that this protein also protects a small subset of cohesin found along the length of the chromosome arms during mitotic prophase. An isoform lacking exon 6 has been shown to play a role in the cohesion of centrioles. Mutations in this gene have been associated with Chronic Atrial and Intestinal Dysrhythmia (CAID) syndrome, characterized by the co-occurrence of Sick Sinus Syndrome (SSS) and Chronic Intestinal Pseudo-obstruction (CIPO) within the first four decades of life. Fibroblast cells from CAID patients exhibited both increased cell proliferation and higher rates of senescence. Pseudogenes of this gene have been found on chromosomes 1 and 7. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-SGOL1 (Ser14)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SGO1
Gene Accession No. Q5FBB7, Q9CXH7
Gene Alias 3300001M08Rik; C81037; CAID; hSgo1; NY-BR-85; serologically defined breast cancer antigen NY-BR-85; SGO; Sgo1; Sgol1; Shugoshin 1; shugoshin 1A protein; shugoshin 1AB protein; shugoshin 1CD protein; shugoshin 1EF protein; shugoshin 1GH protein; shugoshin 1KL protein; shugoshin-like 1; shugoshin-like 1 (S. pombe)
Gene Symbols SGO1
Host Species Rabbit
Immunogen A synthesized peptide derived from human SGO1(Accession Q5FBB7), corresponding to amino acid residues around phosphorylated Ser14.
Purification Method Sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151648, 363174, 72415
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.