Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-SGTA (Ser301) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5105402
Description
Antibody detects endogenous levels of SGTA only when phosphorylated at Ser301.
This gene encodes a protein which is capable of interacting with the major nonstructural protein of parvovirus H-1 and 70-kDa heat shock cognate protein; however, its function is not known. Since this transcript is expressed ubiquitously in various tissues, this protein may serve a housekeeping function.
Specifications
Phospho-SGTA (Ser301) | |
Polyclonal | |
Unconjugated | |
SGTA | |
5330427H01Rik; AI194281; alpha h UBP; alphaSGT; Alpha-SGT; D10Ertd190e; hSGT; protein containing three tetratricopeptide repeats; Sgt; Sgt1; SGTA; small glutamine rich tetratricopeptide repeat containing alpha; small glutamine-rich protein with tetratricopeptide repeats 1; small glutamine-rich tetratricopeptide repeat (TPR) containing protein; small glutamine-rich tetratricopeptide repeat (TPR) containing protein (SGT); small glutamine-rich tetratricopeptide repeat (TPR)-containing, alpha; small glutamine-rich tetratricopeptide repeat-containing protein alpha; Stg; UBP; vpu-binding protein | |
Rabbit | |
Sequential Chromatography | |
RUO | |
52551, 6449, 64667 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O43765, O70593, Q8BJU0 | |
SGTA | |
A synthesized peptide derived from human SGTA(Accession O43765), corresponding to amino acid residues around phosphorylated Ser301. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction