Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-SH2D1B (Tyr127) Polyclonal Antibody, Invitrogen™

Catalog No. PIPA517932
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517932 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517932 Supplier Thermo Scientific Supplier No. PA517932

Goat Polyclonal Antibody

Phospho-SH2D1B (Tyr127) Polyclonal antibody specifically detects Phospho-SH2D1B (Tyr127) in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-SH2D1B (Tyr127)
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Sh2d1b
Gene Accession No. O14796
Gene Alias Eat2; EAT-2; Eat2a; EAT-2A; EWS/FLI1 activated transcript 2; EWS/FLI1-activated transcript 2; RGD1563935; SH2 domain containing 1B; SH2 domain containing 1B1; SH2 domain protein 1B1; SH2 domain-containing molecule EAT2; SH2 domain-containing protein 1B; Sh2d1b; Sh2d1b1
Gene Symbols Sh2d1b
Host Species Goat
Immunogen Synthetic peptide sequence (NSNSDpYVDVLP) corresponding to the internal amino acids of SH2D1B
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117157
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.