Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ phospho-SHC (Tyr239, Tyr240), Rabbit, Unlabeled, Polyclonal,

Catalog No. 07209I Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
07209I 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07209I Supplier MilliporeSigma™ Supplier No. 07209I
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SHC-transforming protein 1 (UniProt P29353; also known as SH2 domain protein C1, SHC-transforming protein 3, SHC-transforming protein A, Src homology 2 domain-containing-transforming protein C1) is encoded by the SHC1 (also known as SHC, SHCA) gene (Gene ID 6464) in human. Originally identified as an SH2-containing proto-oncogene involved in growth factor signaling, Shc proteins function as adaptor molecules recruited by numerous receptors (e.g. integrins, GPCRs, and receptors for growth factors, antigens, cytokines, and hormones) for signling. In addition to the ubiquitously expressed Shc (a.k.a. ShcA by SHC1/SHCA gene), there exist also ShcB (by SHC2/SHCB gene) and ShcC (by SHC3/SHCC gene), which are predominantly expressed in neuronal cells. Alternative splicings result in six ShcA isoforms, including p66Shc, p52Shc, and p46Shc. All ShcA isoforms possess two distinct domains, PTB (a.a. 156-339) and SH2 (a.a. 488-579), that bind phosphotyrosine-containing sequences, as well as a central region (CH1; a.a. 340-487) that contains critical tyrosine phosphorylation sites. The phosphotyrosine-binding (PTB) domain is also referred to as phosphotyrosine-interaction domain or PID.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-SHC (Tyr239, Tyr240)
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene SHC1, SHC, SHCA
Gene Accession No. P29353
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to a target site sequence surrounding phosphorylated Tyr239 and Tyr240 of human spliced isoform p52Shc.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis & Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_892113
Test Specificity Numbering of the target pTyr residues varies depending on the species and spliced isoforms. Target pTyr residues this rabbit polyclonal detects correspond to pTyr349/pTyr350 of human spliced isoform 6 and p66Shc, pTyr239/pTyr240 of human spliced isoform 7 and p52Shc, pTyr194/pTyr195 of isoform p46Shc, and pTyr135/pTyr136 of isoform 5 reported by UniProt (P29353).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.