Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-SHC (Tyr349) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA537737
Description
A suggested positive control for Western blot is HepG2 cells; suggested positive control for ICC/IF is Hela cells.
Skp2 (S-phase Kinase-associated Protein 2) belongs to the family of F-box proteins that interact with the Cyclin A-Cdk2 complex. Skp2 is essential for the G1-S transition in both transformed cells and diploid fibroblasts. Biochemical and genetic experiments have demonstrated that Skp2 is required for the ubiquitination and consequent degradation of p27 in cultured mammalian cells and in vitro reconstitution assays. In normal tissues, Skp2 is expressed in tonsil and placenta. In tumor tissues, Skp2 is expressed widely, in some colon, prostate, pancreas, and skin cancers, with high expression in lung, breast, ovarian, and endometrial cancers. Research suggests that Skp2 plays a role in oncogenesis, particularly in the pathogenesis of lymphomas and breast cancers.
Specifications
Phospho-SHC (Tyr349) | |
Polyclonal | |
Unconjugated | |
SHC1 | |
FLJ26504; OTTHUMP00000035472; p66; P66shc; RP11-307C12.1; SH2 domain protein C1; SHC; SHC (Src homology 2 domain containing) transforming protein 1; SHC (Src homology 2 domain-containing) transforming protein 1; SHC adaptor protein 1; SHC1; SHCA; SHC-transforming protein 1; SHC-transforming protein 3; SHC-transforming protein A; src homology 2 domain-containing transforming protein C1; src homology 2 domain-containing-transforming protein C1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
20416, 6464 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P29353, P98083 | |
SHC1 | |
Peptide sequence around phosphorylation site of tyrosine 349 (H-Q-Y(p)-Y-N) derived from Human Shc1. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction