Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-Ship2 (Tyr1135) Recombinant Rabbit Monoclonal Antibody (Ship2Y1135-1D2), FITC, Invitrogen™
GREENER_CHOICE

Catalog No. PIMA537044
Change view
Click to view available options
Quantity:
10 Tests
100 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537044 100 Tests
PIMA537308 10 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA537044 Supplier Invitrogen™ Supplier No. MA537044
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

The protein encoded by this gene is an SH2-containing 5'-inositol phosphatase that is involved in the regulation of insulin function. The encoded protein also plays a role in the regulation of epidermal growth factor receptor turnover and actin remodeling. Additionally, this gene supports metastatic growth in breast cancer and is a valuable biomarker for breast cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Ship2 (Tyr1135)
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone Ship2Y1135-1D2
Conjugate FITC
Formulation PBS with 0.20% BSA and 0.09% sodium azide
Gene INPPL1
Gene Accession No. O15357, Q6P549
Gene Alias 51C; 51C protein; ablSH3-binding protein; inositol polyphosphate phosphatase like 1; inositol polyphosphate phosphatase-like 1; inositol polyphosphate phosphatase-like protein 1; Inppl1; INPPL-1; NS1; OPSMD; Phosphatidylinositol 3,4,5-trisphosphate 5-phosphatase 2; phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase 2; protein 51C; PTP2C; SH2 domain-containing inositol 5'-phosphatase 2; SH2 domain-containing inositol phosphatase 2; SH2 domain-containing inositol-5'-phosphatase 2; SH2-containing inositol phosphatase 2; SHIP2; SHIP-2; SHP-2; SH-PTP2
Gene Symbols INPPL1
Host Species Rabbit
Immunogen A synthetic phospho-peptide corresponding to residues surrounding Tyr1135 of human phospho Ship2.
Purification Method Protein A/G
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16332, 3636
Target Species Human, Mouse
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.