Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-SHP-1 (Tyr536) Recombinant Rabbit Monoclonal Antibody (Shp1Y536-2A7), FITC, Invitrogen™

Catalog No. PIMA537376
Change view
Click to view available options
Quantity:
10 Tests
100 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537376 10 Tests
PIMA537130 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA537376 Supplier Invitrogen™ Supplier No. MA537376
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

SHP-1 (PTPN6) is a non-receptor protein tyrosine phosphatase that is expressed primarily in hematopoietic cells. The enzyme is composed of two SH2 domains, a tyrosine phosphatase catalytic domain, and a carboxy-terminal regulatory domain. SHP-1 removes phosphates from target proteins to downregulate several tyrosine kinase-regulated pathways. In hematopoietic cells, the amino-terminal SH2 domain of SHP-1 binds to tyrosine phosphorylated erythropoietin receptors (EpoR) to negatively regulate hematopoietic growth. Overexpression of SHP-1 in epithelial cells results in dephosphorylation of the Ros receptor tyrosine kinase and subsequent downregulation of Ros-dependent cell proliferation and transformation. Following ligand binding in myeloid cells, SHP-1 associates with the IL-3R beta chain and downregulates IL-3-induced tyrosine phosphorylation and cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-SHP-1 (Tyr536)
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone Shp1Y536-2A7
Conjugate FITC
Formulation PBS with 0.20% BSA and 0.09% sodium azide
Gene PTPN6
Gene Accession No. P29350
Gene Alias 70Z-SHP; EC 3.1.3.48; hcp; Hcph; hematopoietic cell phosphatase; hematopoietic cell protein-tyrosine phosphatase; HPTP1C; me; motheaten; protein tyrosine phosphatase, non-receptor type 6; protein-tyrosine phosphatase 1C; Protein-tyrosine phosphatase SHP-1; PTN6; Ptp1C; PTP-1C; Ptph6; PTPN6; PTPTY-42; SH2 phosphatase 1; SHP1; Shp-1; SHP-1L; SH-PTP1; Tyrosine-protein phosphatase non-receptor type 6
Gene Symbols PTPN6
Host Species Rabbit
Immunogen A synthetic phospho-peptide corresponding to residues surrounding Tyr536 of human phospho Shp1.
Purification Method Protein A/G
Quantity 10 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5777
Target Species Human
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.