Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-SHP2 (Tyr81) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986694
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6694 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6694 Supplier Bioss Supplier No. BS5638R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is an SH2-containing 5′-inositol phosphatase that is involved in the regulation of insulin function. The encoded protein also plays a role in the regulation of epidermal growth factor receptor turnover and actin remodeling. Additionally, this gene supports metastatic growth in breast cancer and is a valuable biomarker for breast cancer.

Specifications

Antigen Phospho-SHP2 (Tyr81)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene INPPL1
Gene Accession No. O15357
Gene Alias 51C; 51C protein; AblSH3-binding protein; BOS_14953; inositol polyphosphate phosphatase like 1; inositol polyphosphate phosphatase-like 1; Inositol polyphosphate phosphatase-like protein 1; INPPL1; INPPL-1; OPSMD; Phosphatidylinositol 3,4,5-trisphosphate 5-phosphatase 2; phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase 2; Protein 51C; SH2 domain-containing inositol 5′-phosphatase 2; SH2 domain-containing inositol phosphatase 2; SH2 domain-containing inositol-5′-phosphatase 2; SH2-containing inositol phosphatase 2; Ship2; SHIP-2
Gene Symbols INPPL1
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human SHP2 around the phosphorylation site of 81.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3636
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.