Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-SLP76 (Tyr128) Recombinant Rabbit Monoclonal Antibody (SLP76Y128-3F8), APC, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
$253.50 - $639.50
Specifications
| Antigen | Phospho-SLP76 (Tyr128) |
|---|---|
| Clone | SLP76Y128-3F8 |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Conjugate | APC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
PIMA537280
|
Invitrogen™
MA537280 |
10 Tests |
Each for $253.50
|
|
|||||
PIMA537000
![]() |
Invitrogen™
MA537000 |
100 Tests |
Each for $639.50
|
|
|||||
Description
SLP-76 was originally identified as a substrate of the ZAP-70 protein tyrosine kinase following T cell receptor (TCR) ligation in the leukemic T cell line Jurkat. The SLP-76 locus has been localized to human chromosome 5q33 and the gene structure has been partially characterized in mice. The human and murine cDNAs both encode 533 amino acid proteins that are 72% identical and comprised of three modular domains. The NH2-terminus contains an acidic region that includes a PEST domain and several tyrosine residues which are phosphorylated following TCR ligation. SLP-76 also contains a central proline-rich domain and a COOH-terminal SH2 domain. A number of additional proteins have been identified that associate with SLP-76 both constitutively and inducibly following receptor ligation, supporting the notion that SLP-76 functions as an adaptor or scaffold protein. Studies using SLP-76 deficient T cell lines or mice have provided strong evidence that SLP-76 plays a positive role in promoting T cell development and activation as well as mast cell and platelet function.Specifications
| Phospho-SLP76 (Tyr128) | |
| Flow Cytometry | |
| APC | |
| Rabbit | |
| Human, Mouse | |
| Q13094, Q60787 | |
| 16822, 3937 | |
| A synthetic phospho-peptide corresponding to residues surrounding Tyr128 of human phospho SLP-76. | |
| Primary | |
| 4°C | |
| LCP2 |
| SLP76Y128-3F8 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.20% BSA and 0.09% sodium azide | |
| 76 kDa tyrosine phosphoprotein; AI323664; BB161688; Lcp2; Lymphocyte cytosolic protein 2; lymphocyte cytosolic protein 2 (SH2 domain containing leukocyte protein of 76kDa); lymphocyte cytosolic protein 2 (SH2 domain-containing leukocyte protein of 76kD); SH2 domain-containing leukocyte protein of 76 kDa; SH2 domain-containing leukocyte protein of 76kD; SLP76; SLP-76; SLP-76 tyrosine phosphoprotein; twm | |
| LCP2 | |
| IgG κ | |
| Protein A/G | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title