Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-SMAD1 (Ser214) Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5105798
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105798 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105798 Supplier Thermo Scientific Supplier No. PA5105798

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of Smad1 only when phosphorylated at Ser214.

The protein encoded by this gene belongs to the SMAD, a family of proteins similar to the gene products of the Drosophila gene 'mothers against decapentaplegic' (Mad) and the C. elegans gene Sma. SMAD proteins are signal transducers and transcriptional modulators that mediate multiple signaling pathways. This protein mediates the signals of the bone morphogenetic proteins (BMPs), which are involved in a range of biological activities including cell growth, apoptosis, morphogenesis, development and immune responses. In response to BMP ligands, this protein can be phosphorylated and activated by the BMP receptor kinase. The phosphorylated form of this protein forms a complex with SMAD4, which is important for its function in the transcription regulation. This protein is a target for SMAD-specific E3 ubiquitin ligases, such as SMURF1 and SMURF2, and undergoes ubiquitination and proteasome-mediated degradation. Alternatively spliced transcript variants encoding the same protein have been observed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-SMAD1 (Ser214)
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SMAD1
Gene Accession No. P70340, P97588, Q15797
Gene Alias AI528653, BSP1, BSP-1, dwarfin-A, Dwf-A, DWFC, hSMAD1, JV41, JV4-1, JV5-1, MAD (mothers against decapentaplegic, Drosophila) homolog 1, MAD homolog 1, MAD homolog1 (mothers against decapentaplegic, Drosophila), MAD, mothers against decapentaplegic homolog 1, Madh1, MADH5, MADR1, Mad-related protein 1, Mlp1, mMad1, mothers against decapentaplegic homolog 1, mothers against DPP homolog 1, mothers-against-DPP-related 1, Mothers-against-DPP-related-1, MusMLP, OTTHUMP00000220456, SMAD, Smad 1, SMAD family member 1, SMAD, mothers against DPP homolog 1, Smad1, SMAD5, TGF-beta signaling protein 1, transforming growth factor-beta signaling protein 1, Transforming growth factor-beta-signaling protein 1
Gene Symbols SMAD1
Host Species Rabbit
Immunogen A synthesized peptide derived from human SMAD1(Accession Q15797), corresponding to amino acid residues around phosphorylated Ser214.
Purification Method sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17125, 25671, 4086
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.