Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ phospho Smad1/Smad5/Smad8 (Ser463/465), Rabbit, Unlabeled, Polyclonal,

Catalog No. AB3848IMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
AB3848IMI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3848IMI Supplier MilliporeSigma™ Supplier No. AB3848I
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Mothers against decapentaplegic homolog 1 (UniProt Q15797; also known as BSP-1, hSMAD1, JV4-1, MAD homolog 1, Mothers against DPP homolog 1, Mad-related protein 1, SMAD 1, SMAD family member 1, Smad1, Transforming growth factor-beta-signaling protein 1) is encoded by the SMAD1 (also known as BSP1, MADH1, MADR1) gene (Gene ID 4086) in human. Mammalian SMADs constitute a family of transcription factors that are subdivided into three groups. SMAD1, 2, 3, 5, and 9 (a.k.a. SMAD8) are receptor-regulated SMADs or R-SMADs. SMAD4 is the only known mammalian common mediator SMAD or Co-SMAD and it mediates both the TGF- and BMP signalling pathways. SMAD6 and SMAD7 are inhibitory SMADs or I-SMADs that block the activation of R-SMADs by Co-SMAD via competitive binding. SMADs share a similar structure feature consisting of the conserved N-terminal Mad-homology 1 (MH1; a.a. 12-136 of hSMAD1, a.a. 13-137 of hSMAD2, a.a. 16-140 of hSMAD8) and C-terminal MH2 (a.a. 271-465 of hSMAD1 and hSMAD2, a.a. 273-467 of hSMAD8) domains joined by a non-conserved linker region among SMADs. MH1 domain in R-SMADs and Co-SMADs contains N-terminus nuclear localization signals (NLS), while SMAD4 has nuclear export signals (NES) in this domain. The MH2 domain is responsible for gene transactivation as well as for mediating SMAD-receptor, SMAD-SMAD, SMAD-coactivators and SMAD-corepressors interactions. A TGF- ligand (TGF- for SMAD2/3 and BMP for SMAD1/5/8) initiates signalling by binding and engaging type I and type II receptor serine/threonine kinases on the cell surface, allowing T RII-dependent phosphorylation of T RI kinase domain, which then instigates signalling by phosphorylating the last two serine residues within the conserved C-terminal end Ser-Ser-X-Ser (SSXS) motif of SMAD proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho Smad1/Smad5/Smad8 (Ser463/465)
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibdoy in buffer containing 0.1M Tris-Glycine (pH 7.4) 150mM NaCl with 0.05% Sodium Azide.
Gene SMAD1, BSP1, MADH1, MADR1
Gene Accession No. Q15797
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human SMAD1 C-terminal end sequence phosphorylated on the last two serine residues (S463/S465).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_005891
Test Specificity This rabbit polyclonal antibody recognizes SMAD1/5/8 phosphorylated on their last two serine residues within the C-terminal end Ser-Ser-X-Ser (SSXS) motif. Target serine residues are present in human/mouse SMAD1 (S463/S465), rat SMAD1 (S466/S468), human/mouse/rat SMAD5 (S463/S465), human SMAD9 (S465/S467), mouse SMAD9 (S428/S430), and rat SMAD9 (S432/S434), but absent in human SMAD1 spliced isoform 2. SMAD9 is also known as SMAD8. Target bands detection was blocked by the immunogen phosphopeptide, but not by the corresponding non-phosphopeptide.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.